Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTTAATGCTTTGAGGGTTTCT[A/C]GAAATACAGAAAGTCATCAAGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf194 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1orf194 - chromosome 1 open reading frame 194 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA2 - small Cajal body-specific RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM167B - transmembrane protein 167B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322248.1 | 1947 | UTR 3 | NP_001309177.1 | |||
NM_020141.3 | 1947 | UTR 3 | NP_064526.1 |