Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACCTGCTCAACCTGGCCATCTC[C/T]GACCTGTTTTTCCTTCTTACTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601373 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCR5 - C-C motif chemokine receptor 5 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000579.3 | 582 | Silent Mutation | TCC,TCT | S,S 75 | NP_000570.1 | |
NM_001100168.1 | 582 | Silent Mutation | TCC,TCT | S,S 75 | NP_001093638.1 |
LOC102724297 - uncharacterized LOC102724297 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |