Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTCTGGGGCCCGGCCAGTGGCTG[C/G]AGCCAGGCTCTGCAGCGCCTCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 176873 | ||||||||||||||||||||
Literature Links: |
CDK11B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK11B - cyclin dependent kinase 11B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35E2B - solute carrier family 35 member E2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110781.2 | 2068 | Missense Mutation | CCA,GCA | P,A 383 | NP_001104251.1 | |
NM_001290264.1 | 2068 | Missense Mutation | CCA,GCA | P,A 383 | NP_001277193.1 | |
XM_011542072.1 | 2068 | Missense Mutation | TCC,TGC | S,C 300 | XP_011540374.1 |