Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCGGGTACTTACACACTGGCA[G/T]AGCTCACAGATGAGAAACGCCCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612316 MIM: 616101 | ||||||||||||||||||||
Literature Links: |
ATAD3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATAD3A - ATPase family, AAA domain containing 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSU72 - SSU72 homolog, RNA polymerase II CTD phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014188.2 | 799 | Silent Mutation | CTA,CTC | L,L 158 | NP_054907.1 |
TMEM240 - transmembrane protein 240 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |