Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCACCCCTGCTTTTCTTTCTTC[A/G]TCCCACTAATATTGTCTTAGATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609611 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANP32E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANP32E - acidic nuclear phosphoprotein 32 family member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136478.3 | 1986 | UTR 3 | NP_001129950.1 | |||
NM_001136479.2 | 1986 | UTR 3 | NP_001129951.1 | |||
NM_001280559.1 | 1986 | UTR 3 | NP_001267488.1 | |||
NM_001280560.1 | 1986 | UTR 3 | NP_001267489.1 | |||
NM_030920.4 | 1986 | UTR 3 | NP_112182.1 | |||
XM_005245513.3 | 1986 | UTR 3 | XP_005245570.1 | |||
XM_005245514.3 | 1986 | UTR 3 | XP_005245571.1 | |||
XM_017002418.1 | 1986 | UTR 3 | XP_016857907.1 |