Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTTACAAAACTCAGACCACTCA[C/G]CAGAAAAAAATCGGCATTTATATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615255 | ||||||||||||||||||||
Literature Links: |
C1orf112 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf112 - chromosome 1 open reading frame 112 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL18 - methyltransferase like 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320199.1 | 1062 | Missense Mutation | GCT,GGT | A,G 268 | NP_001307128.1 | |
NM_001320201.1 | 1062 | Missense Mutation | GCT,GGT | A,G 268 | NP_001307130.1 | |
NM_033418.3 | 1062 | Missense Mutation | GCT,GGT | A,G 268 | NP_219486.1 | |
XM_006711627.3 | 1062 | Missense Mutation | GCT,GGT | A,G 268 | XP_006711690.1 |