Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAGGATACATTTTCAGATAAC[A/G]GCAAGGTAAATAATGAGGTAAGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SETDB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SETDB1 - SET domain bifurcated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145415.1 | Intron | NP_001138887.1 | ||||
NM_001243491.1 | Intron | NP_001230420.1 | ||||
NM_012432.3 | Intron | NP_036564.3 | ||||
XM_005245641.3 | Intron | XP_005245698.1 | ||||
XM_017002953.1 | Intron | XP_016858442.1 | ||||
XM_017002954.1 | Intron | XP_016858443.1 | ||||
XM_017002955.1 | Intron | XP_016858444.1 |