Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGATTCCATCATCCCAAATTTC[C/T]ATCTCTCCAGTTCTTTGGTCCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605834 MIM: 600607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMOD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMOD4 - tropomodulin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS72 - vacuolar protein sorting 72 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271087.1 | Intron | NP_001258016.1 | ||||
NM_001271088.1 | Intron | NP_001258017.1 | ||||
NM_005997.2 | Intron | NP_005988.1 | ||||
XM_017002205.1 | Intron | XP_016857694.1 |