Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACTGAGTTCCACACCAAACTTG[A/G]TACCATCACAATTACATTTTAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611063 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM20B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM20B - family with sequence similarity 20 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324310.1 | Intron | NP_001311239.1 | ||||
NM_001324311.1 | Intron | NP_001311240.1 | ||||
NM_014864.3 | Intron | NP_055679.1 | ||||
XM_017003001.1 | Intron | XP_016858490.1 | ||||
XM_017003002.1 | Intron | XP_016858491.1 |