Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTAAATTATTTTGAAGAGAAAT[G/T]ATGAAAAACCATATTCAACTGAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOR1AIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOR1AIP2 - torsin 1A interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199260.1 | 3596 | UTR 3 | NP_001186189.1 | |||
NM_022347.3 | 3596 | Intron | NP_071742.1 | |||
NM_145034.4 | 3596 | UTR 3 | NP_659471.1 | |||
XM_005244939.4 | 3596 | UTR 3 | XP_005244996.1 | |||
XM_017000474.1 | 3596 | UTR 3 | XP_016855963.1 | |||
XM_017000475.1 | 3596 | UTR 3 | XP_016855964.1 | |||
XM_017000476.1 | 3596 | UTR 3 | XP_016855965.1 | |||
XM_017000477.1 | 3596 | Intron | XP_016855966.1 | |||
XM_017000478.1 | 3596 | Intron | XP_016855967.1 |