Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTCCTCTTTAGGTGGTTGGTA[G/T]TTTGAACTGTTTTAGCACTTTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SWT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SWT1 - SWT1, RNA endoribonuclease homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105518.1 | Intron | NP_001098988.1 | ||||
NM_017673.6 | Intron | NP_060143.4 | ||||
XM_005245271.1 | Intron | XP_005245328.1 | ||||
XM_005245273.1 | Intron | XP_005245330.1 | ||||
XM_011509657.1 | Intron | XP_011507959.1 | ||||
XM_017001527.1 | Intron | XP_016857016.1 | ||||
XM_017001528.1 | Intron | XP_016857017.1 | ||||
XM_017001529.1 | Intron | XP_016857018.1 | ||||
XM_017001530.1 | Intron | XP_016857019.1 |