Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGACCAGCAGCAGATCCAGTGCC[C/G]CCTGCCGCTCCAACAGTGCTGCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613260 | ||||||||||||||||||||
Literature Links: |
KPRP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KPRP - keratinocyte proline rich protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025231.1 | 128 | Missense Mutation | CCC,CGC | P,R 10 | NP_001020402.1 | |
XM_011509571.1 | 128 | Missense Mutation | CCC,CGC | P,R 10 | XP_011507873.1 |