Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCCAGAAATCTGCCCTCTCACA[A/G]GAATTCCTCTTCTGACCTCTGATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612607 MIM: 612608 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCE1E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCE1E - late cornified envelope 1E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCE1F - late cornified envelope 1F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178354.2 | 516 | UTR 3 | NP_848131.1 |