Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGGAGGAGGGAGTGGCCAGCA[C/G]TCTGGAGGCTGCTGCTAAAGTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612604 MIM: 612605 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCE1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCE1B - late cornified envelope 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178349.1 | 815 | Missense Mutation | CAC,CAG | H,Q 113 | NP_848126.1 |
LCE1C - late cornified envelope 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |