Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAATGCAGTATAAATTCACAAA[G/T]GAAATGCTACAGCAGATGGTGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605337 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFHR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFHR4 - complement factor H related 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201550.2 | Intron | NP_001188479.1 | ||||
NM_001201551.1 | Intron | NP_001188480.1 | ||||
NM_006684.4 | Intron | NP_006675.2 | ||||
XM_006711129.3 | Intron | XP_006711192.1 | ||||
XM_017000110.1 | Intron | XP_016855599.1 | ||||
XM_017000111.1 | Intron | XP_016855600.1 | ||||
XM_017000112.1 | Intron | XP_016855601.1 | ||||
XM_017000113.1 | Intron | XP_016855602.1 | ||||
XM_017000114.1 | Intron | XP_016855603.1 |