Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGCCGTTTTTGAGAAATTCCC[A/G]GGTATTTTGTGGGCCTTGAAGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614206 MIM: 176940 MIM: 601989 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHTOP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHTOP - chromatin target of PRMT1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206612.1 | Intron | NP_001193541.1 | ||||
NM_001244664.1 | Intron | NP_001231593.1 | ||||
NM_001317077.1 | Intron | NP_001304006.1 | ||||
NM_015607.3 | Intron | NP_056422.2 | ||||
XM_017000993.1 | Intron | XP_016856482.1 |
S100A1 - S100 calcium binding protein A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S100A13 - S100 calcium binding protein A13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |