Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTCTGAGATCCAAAAAACCCAG[C/T]CTTCCCCCCTCCTCATCTTGGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123876 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSRP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSRP1 - cysteine and glycine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193570.1 | 1223 | UTR 3 | NP_001180499.1 | |||
NM_001193571.1 | 1223 | UTR 3 | NP_001180500.1 | |||
NM_001193572.1 | 1223 | UTR 3 | NP_001180501.1 | |||
NM_004078.2 | 1223 | UTR 3 | NP_004069.1 |