Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGCCAGCTCGGGCAGGGCCAG[C/T]ATGGGCATCTCCTGAGACCCCACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603768 MIM: 600104 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R12B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R12B - protein phosphatase 1 regulatory subunit 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT2 - synaptotagmin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136504.1 | 6928 | UTR 3 | NP_001129976.1 | |||
NM_177402.4 | 6928 | UTR 3 | NP_796376.2 | |||
XM_011509192.2 | 6928 | UTR 3 | XP_011507494.1 | |||
XM_017000309.1 | 6928 | UTR 3 | XP_016855798.1 | |||
XM_017000310.1 | 6928 | UTR 3 | XP_016855799.1 | |||
XM_017000311.1 | 6928 | UTR 3 | XP_016855800.1 | |||
XM_017000312.1 | 6928 | UTR 3 | XP_016855801.1 | |||
XM_017000313.1 | 6928 | UTR 3 | XP_016855802.1 |