Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAGATGAGCCGTGGCTGTGGGG[A/G]GCAGTAGAAGGAATAATAGAGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607945 | ||||||||||||||||||||
Literature Links: |
ADIPOR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADIPOR1 - adiponectin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290553.1 | 997 | Intron | NP_001277482.1 | |||
NM_001290557.1 | 997 | Missense Mutation | CCC,TCC | P,S 231 | NP_001277486.1 | |
NM_001290629.1 | 997 | Intron | NP_001277558.1 | |||
NM_015999.5 | 997 | Missense Mutation | CCC,TCC | P,S 231 | NP_057083.2 | |
XM_017001407.1 | 997 | Intron | XP_016856896.1 |