Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGGTGTCTCCAGCCTCCACCTC[C/T]TTCTCTGTGAGGTGGAAATTCTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605127 MIM: 601914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPTC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OPTC - opticin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014359.3 | Intron | NP_055174.1 | ||||
XM_011509406.2 | Intron | XP_011507708.1 |
PRELP - proline and arginine rich end leucine rich repeat protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |