Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAGCCCTGCAAACCCCCGCTCA[A/G]GTCTAGAGGACATGGAGCTGCCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 169190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK18 - cyclin dependent kinase 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002596.3 | Intron | NP_002587.2 | ||||
NM_212502.2 | Intron | NP_997667.1 | ||||
NM_212503.2 | Intron | NP_997668.1 | ||||
XM_011509602.2 | Intron | XP_011507904.1 | ||||
XM_017001423.1 | Intron | XP_016856912.1 |