Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGCTACCTGGTCCCTACCATCG[A/T]GGGGTGAGTGGAACTTAGAGAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSTO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSTO1 - misato 1, mitochondrial distribution and morphology regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256532.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | NP_001243461.1 | |
NM_001256533.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | NP_001243462.1 | |
NM_018116.3 | 897 | Silent Mutation | CGA,CGT | R,R 270 | NP_060586.2 | |
XM_006711421.1 | 897 | Silent Mutation | CGA,CGT | R,R 215 | XP_006711484.1 | |
XM_006711423.2 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_006711486.1 | |
XM_011509695.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011507997.1 | |
XM_011509696.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011507998.1 | |
XM_011509697.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011507999.1 | |
XM_011509698.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011508000.1 | |
XM_011509699.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011508001.1 | |
XM_011509700.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011508002.1 | |
XM_011509701.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011508003.1 | |
XM_011509702.1 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011508004.1 | |
XM_011509703.1 | 897 | Silent Mutation | CGA,CGT | R,R 215 | XP_011508005.1 | |
XM_011509704.1 | 897 | Silent Mutation | CGA,CGT | R,R 215 | XP_011508006.1 | |
XM_011509705.1 | 897 | Silent Mutation | CGA,CGT | R,R 215 | XP_011508007.1 | |
XM_011509706.1 | 897 | Silent Mutation | CGA,CGT | R,R 93 | XP_011508008.1 | |
XM_011509707.1 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_011508009.1 | |
XM_011509708.1 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_011508010.1 | |
XM_011509709.2 | 897 | Silent Mutation | CGA,CGT | R,R 270 | XP_011508011.1 | |
XM_017001603.1 | 897 | Silent Mutation | CGA,CGT | R,R 93 | XP_016857092.1 | |
XM_017001604.1 | 897 | Silent Mutation | CGA,CGT | R,R 93 | XP_016857093.1 | |
XM_017001605.1 | 897 | Silent Mutation | CGA,CGT | R,R 93 | XP_016857094.1 | |
XM_017001606.1 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_016857095.1 | |
XM_017001607.1 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_016857096.1 | |
XM_017001608.1 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_016857097.1 | |
XM_017001609.1 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_016857098.1 | |
XM_017001610.1 | 897 | Silent Mutation | CGA,CGT | R,R 89 | XP_016857099.1 | |
XM_017001611.1 | 897 | Silent Mutation | CGA,CGT | R,R 89 | XP_016857100.1 | |
XM_017001612.1 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_016857101.1 | |
XM_017001613.1 | 897 | Silent Mutation | CGA,CGT | R,R 89 | XP_016857102.1 | |
XM_017001614.1 | 897 | Silent Mutation | CGA,CGT | R,R 92 | XP_016857103.1 | |
XM_017001615.1 | 897 | Silent Mutation | CGA,CGT | R,R 89 | XP_016857104.1 | |
XM_017001616.1 | 897 | Silent Mutation | CGA,CGT | R,R 93 | XP_016857105.1 |