Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATAATATGGTGGTTGAGGAGGG[A/G]CACTACTTATAGCAGAGGGTTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0907 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0907 - KIAA0907 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014949.3 | 1112 | Missense Mutation | GCC,GTC | A,V 348 | NP_055764.2 |
SCARNA4 - small Cajal body-specific RNA 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA80E - small nucleolar RNA, H/ACA box 80E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |