Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGGGATGAAGGATGTGACCAG[A/G]ACCAGAGTGGCTTGTGATCTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607079 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHBG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RHBG - Rh family B glycoprotein (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256395.1 | Intron | NP_001243324.1 | ||||
NM_001256396.1 | Intron | NP_001243325.1 | ||||
NM_020407.4 | Intron | NP_065140.3 | ||||
XM_011509799.2 | Intron | XP_011508101.1 | ||||
XM_011509800.2 | Intron | XP_011508102.1 | ||||
XM_017001857.1 | Intron | XP_016857346.1 | ||||
XM_017001858.1 | Intron | XP_016857347.1 | ||||
XM_017001859.1 | Intron | XP_016857348.1 |