Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCTCCAAGGTGGCTGCCTCCTT[C/T]TCCTCTCCTTCAGGGTTTTCTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600339 MIM: 611836 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDGF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDGF - hepatoma-derived growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126050.1 | 645 | Silent Mutation | GAA,GAG | E,E 195 | NP_001119522.1 | |
NM_001126051.1 | 645 | Silent Mutation | GAA,GAG | E,E 172 | NP_001119523.1 | |
NM_001319186.1 | 645 | Silent Mutation | GAA,GAG | E,E 202 | NP_001306115.1 | |
NM_001319187.1 | 645 | Silent Mutation | GAA,GAG | E,E 147 | NP_001306116.1 | |
NM_001319188.1 | 645 | Silent Mutation | GAA,GAG | E,E 147 | NP_001306117.1 | |
NM_004494.2 | 645 | Silent Mutation | GAA,GAG | E,E 179 | NP_004485.1 | |
XM_011509453.2 | 645 | Silent Mutation | GAA,GAG | E,E 147 | XP_011507755.2 | |
XM_011509454.2 | 645 | Silent Mutation | GAA,GAG | E,E 147 | XP_011507756.1 | |
XM_011509455.2 | 645 | Silent Mutation | GAA,GAG | E,E 147 | XP_011507757.1 |
MRPL24 - mitochondrial ribosomal protein L24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RRNAD1 - ribosomal RNA adenine dimethylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |