Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCCCACAAACTGATTTCCCAGA[C/T]GTTGTTGGTGAAGATATAAATTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606508 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCRL1 - Fc receptor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159397.1 | 2734 | UTR 3 | NP_001152869.1 | |||
NM_001159398.1 | 2734 | UTR 3 | NP_001152870.1 | |||
NM_052938.4 | 2734 | UTR 3 | NP_443170.1 | |||
XM_005244866.2 | 2734 | UTR 3 | XP_005244923.1 | |||
XM_005244867.2 | 2734 | UTR 3 | XP_005244924.1 | |||
XM_005244869.3 | 2734 | UTR 3 | XP_005244926.1 | |||
XM_011509125.1 | 2734 | UTR 3 | XP_011507427.1 | |||
XM_011509126.1 | 2734 | UTR 3 | XP_011507428.1 | |||
XM_011509127.2 | 2734 | UTR 3 | XP_011507429.1 | |||
XM_011509128.1 | 2734 | UTR 3 | XP_011507430.1 | |||
XM_011509129.1 | 2734 | UTR 3 | XP_011507431.1 | |||
XM_011509130.1 | 2734 | UTR 3 | XP_011507432.1 | |||
XM_011509131.1 | 2734 | Intron | XP_011507433.1 | |||
XM_011509132.2 | 2734 | Intron | XP_011507434.1 | |||
XM_011509133.2 | 2734 | UTR 3 | XP_011507435.1 | |||
XM_011509134.2 | 2734 | UTR 3 | XP_011507436.1 | |||
XM_011509135.2 | 2734 | UTR 3 | XP_011507437.1 | |||
XM_011509136.1 | 2734 | Intron | XP_011507438.1 | |||
XM_011509137.2 | 2734 | UTR 3 | XP_011507439.1 | |||
XM_017000227.1 | 2734 | UTR 3 | XP_016855716.1 |