Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTACACTACGCCTTATGCGGGC[A/G]AGGTATGTGGATAACAGGGAAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCEA3 - transcription elongation factor A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003196.2 | Intron | NP_003187.1 | ||||
XM_006710864.1 | Intron | XP_006710927.1 | ||||
XM_011542053.2 | Intron | XP_011540355.1 | ||||
XM_017002200.1 | Intron | XP_016857689.1 | ||||
XM_017002201.1 | Intron | XP_016857690.1 | ||||
XM_017002202.1 | Intron | XP_016857691.1 | ||||
XM_017002203.1 | Intron | XP_016857692.1 | ||||
XM_017002204.1 | Intron | XP_016857693.1 |