Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGCACATCTCCCACTTGTGAT[C/T]TGGCAAGCTCTGTGGCTTGAATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUCA1 - fucosidase, alpha-L- 1, tissue | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000147.4 | 536 | Missense Mutation | AAT,GAT | N,D 290 | NP_000138.2 | |
XM_005245821.2 | 536 | Missense Mutation | AAT,GAT | N,D 165 | XP_005245878.1 | |
XM_011541167.2 | 536 | Missense Mutation | AAT,GAT | N,D 79 | XP_011539469.1 | |
XM_017000905.1 | 536 | Missense Mutation | AAT,GAT | N,D 189 | XP_016856394.1 |