Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTGGAAATCAGACGACCCCAAT[A/C]CCATCGAGGCTTTCTATCTTCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608317 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRHL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRHL3 - grainyhead like transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195010.1 | 84 | Intron | NP_001181939.1 | |||
NM_021180.3 | 84 | Intron | NP_067003.2 | |||
NM_198173.2 | 84 | Intron | NP_937816.1 | |||
NM_198174.2 | 84 | Intron | NP_937817.3 | |||
XM_011541869.1 | 84 | UTR 5 | XP_011540171.1 | |||
XM_011541870.2 | 84 | Intron | XP_011540172.1 |