Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGTATAGGGGAGGGCTGGAGAG[A/C]TCCAGATGCTACAGATGTCCCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601738 MIM: 609617 MIM: 606131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXTL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXTL1 - exostosin like glycosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC30A2 - solute carrier family 30 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004434.2 | Intron | NP_001004434.1 | ||||
NM_032513.4 | Intron | NP_115902.1 |
TRIM63 - tripartite motif containing 63 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |