Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGAACCCCCTTACCTGGACATGC[C/T]TCTGAGCGGTTGTAGATGGAGCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604973 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD164L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD164L2 - CD164 molecule like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005245868.3 | 394 | Silent Mutation | GAA,GAG | E,E 106 | XP_005245925.2 | |
XM_011541441.1 | 394 | Silent Mutation | GAA,GAG | E,E 106 | XP_011539743.1 |
FCN3 - ficolin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |