Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGGGCACCAATATTACCTATGA[C/T]GACGCTGCTGCCACCAGCCCCTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147572 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFI6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFI6 - interferon alpha inducible protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002038.3 | 454 | Missense Mutation | ATC,GTC | I,V 107 | NP_002029.3 | |
NM_022872.2 | 454 | Missense Mutation | ATC,GTC | I,V 111 | NP_075010.1 | |
NM_022873.2 | 454 | Missense Mutation | ATC,GTC | I,V 115 | NP_075011.1 |