Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAATTTTTTCCCTGCATATCCAA[C/T]AGTGTCATGTATCTCTTTTCCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMEB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GMEB1 - glucocorticoid modulatory element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319674.1 | Intron | NP_001306603.1 | ||||
NM_006582.3 | Intron | NP_006573.2 | ||||
NM_024482.2 | Intron | NP_077808.1 | ||||
XM_011540518.2 | Intron | XP_011538820.1 | ||||
XM_011540519.2 | Intron | XP_011538821.1 | ||||
XM_011540521.2 | Intron | XP_011538823.1 | ||||
XM_011540522.2 | Intron | XP_011538824.1 | ||||
XM_017000087.1 | Intron | XP_016855576.1 |