Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTCGTCGATGAAGGTGCATTC[A/G]AGGCCCAGGGGCTCCAGGAGGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607935 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PADI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PADI2 - peptidyl arginine deiminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007365.2 | 1137 | Silent Mutation | CTC,CTT | L,L 627 | NP_031391.2 | |
XM_017000148.1 | 1137 | Silent Mutation | CTC,CTT | L,L 312 | XP_016855637.1 |