Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCTGCAAATTGGACTCAGGCC[C/T]CTGGGACATTTCCCAAGGCCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605611 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3BP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH3BP4 - SH3 domain binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014521.2 | Intron | NP_055336.1 | ||||
XM_011510891.2 | Intron | XP_011509193.1 | ||||
XM_011510892.1 | Intron | XP_011509194.1 | ||||
XM_011510893.1 | Intron | XP_011509195.1 | ||||
XM_011510894.2 | Intron | XP_011509196.1 |