Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCACCACCCTGACCTTGGATGG[A/G]TGGCAAGACTTGCCACCCCCACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610376 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACKR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACKR3 - atypical chemokine receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020311.2 | Intron | NP_064707.1 | ||||
XM_005246097.2 | Intron | XP_005246154.1 | ||||
XM_005246098.3 | Intron | XP_005246155.1 | ||||
XM_017004516.1 | Intron | XP_016860005.1 |