Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGCACCATCTACCAGGCCTTTA[C/T]ACGAACTGAACTCTCCCCATTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL6A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL6A3 - collagen type VI alpha 3 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004369.3 | Intron | NP_004360.2 | ||||
NM_057164.4 | Intron | NP_476505.3 | ||||
NM_057165.4 | Intron | NP_476506.3 | ||||
NM_057166.4 | Intron | NP_476507.3 | ||||
NM_057167.3 | Intron | NP_476508.2 | ||||
XM_005246065.1 | Intron | XP_005246122.1 | ||||
XM_005246066.1 | Intron | XP_005246123.1 | ||||
XM_006712253.1 | Intron | XP_006712316.1 | ||||
XM_011510574.1 | Intron | XP_011508876.1 | ||||
XM_017003303.1 | Intron | XP_016858792.1 | ||||
XM_017003304.1 | Intron | XP_016858793.1 |