Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTTGAGAGTTGACTGTTTAATT[C/T]ATTTATTTCTAGCTTTATTTAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM44 - RNA binding motif protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080504.2 | Intron | NP_001073973.2 | ||||
XM_005246075.1 | Intron | XP_005246132.1 | ||||
XM_011511152.2 | Intron | XP_011509454.1 | ||||
XM_011511153.2 | Intron | XP_011509455.1 | ||||
XM_011511154.2 | Intron | XP_011509456.1 | ||||
XM_011511155.1 | Intron | XP_011509457.1 | ||||
XM_017004054.1 | Intron | XP_016859543.1 | ||||
XM_017004055.1 | Intron | XP_016859544.1 | ||||
XM_017004056.1 | Intron | XP_016859545.1 | ||||
XM_017004057.1 | Intron | XP_016859546.1 | ||||
XM_017004058.1 | Intron | XP_016859547.1 |