Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCGAGTCCCGTGAGCTTACTGCCA[C/T]GTCGATGGCTGCGGGCAGGCCGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610331 MIM: 603426 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HES6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HES6 - hes family bHLH transcription factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PER2 - period circadian clock 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022817.2 | 3788 | Missense Mutation | ATG,GTG | M,V 1205 | NP_073728.1 | |
XM_005246111.4 | 3788 | Missense Mutation | ATG,GTG | M,V 1205 | XP_005246168.1 | |
XM_006712824.3 | 3788 | Missense Mutation | ATG,GTG | M,V 1205 | XP_006712887.1 |