Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAAACAGGGCTAACAATGCATA[A/G]GGTAGTGTATACCAGATGATGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608025 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBAS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NBAS - neuroblastoma amplified sequence | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015909.3 | Intron | NP_056993.2 | ||||
XM_011510357.2 | Intron | XP_011508659.1 | ||||
XM_011510358.2 | Intron | XP_011508660.1 | ||||
XM_011510359.2 | Intron | XP_011508661.1 | ||||
XM_011510360.2 | Intron | XP_011508662.1 | ||||
XM_011510361.2 | Intron | XP_011508663.1 | ||||
XM_017004317.1 | Intron | XP_016859806.1 |