Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAAATAATTTCAAGGAACAGTA[A/C]TCACTGTTATTTTAGAGTATTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611787 MIM: 607810 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMPK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CMPK2 - cytidine/uridine monophosphate kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSAD2 - radical S-adenosyl methionine domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080657.4 | Intron | NP_542388.2 | ||||
XM_011510414.2 | Intron | XP_011508716.1 | ||||
XM_011510415.2 | Intron | XP_011508717.1 |