Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGCAAACTCTGTCGCATCTGTT[A/C]TGGGTCAGAGCTTTGGAATGTTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610318 | ||||||||||||||||||||
Literature Links: |
COBLL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COBLL1 - cordon-bleu WH2 repeat protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278458.1 | 3571 | Nonsense Mutation | GAA,TAA | E,* 1184 | NP_001265387.1 | |
NM_001278460.1 | 3571 | Nonsense Mutation | GAA,TAA | E,* 1125 | NP_001265389.1 | |
NM_001278461.1 | 3571 | Nonsense Mutation | GAA,TAA | E,* 1079 | NP_001265390.1 | |
NM_014900.4 | 3571 | Nonsense Mutation | GAA,TAA | E,* 1117 | NP_055715.3 |
SNORA70F - small nucleolar RNA, H/ACA box 70F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |