Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGATTTGTGTAAGGAATATACT[A/G]CTGTTCTTGCCATTTCCAAAGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109090 MIM: 613831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL5 - methyltransferase like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293186.1 | 648 | Missense Mutation | GCA,GTA | A,V 152 | NP_001280115.1 | |
NM_001293187.1 | 648 | Missense Mutation | GCA,GTA | A,V 152 | NP_001280116.1 | |
NM_014168.3 | 648 | Missense Mutation | GCA,GTA | A,V 152 | NP_054887.2 | |
XM_011511023.1 | 648 | Missense Mutation | GCA,GTA | A,V 152 | XP_011509325.1 |
SSB - Sjogren syndrome antigen B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBR3 - ubiquitin protein ligase E3 component n-recognin 3 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |