Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTCAGCTGTACAGTTTGGGAA[A/G]ATACCTACACACCCGGAGAACAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GORASP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GORASP2 - golgi reassembly stacking protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201428.1 | 317 | Intron | NP_001188357.1 | |||
NM_015530.4 | 317 | UTR 5 | NP_056345.3 | |||
XM_006712408.2 | 317 | Intron | XP_006712471.1 |