Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGGCAGGTGCAGAGTAACATTC[G/T]TTTTTTGTTCTTGAAAGCAAAGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAPGEF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAPGEF4 - Rap guanine nucleotide exchange factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100397.1 | Intron | NP_001093867.1 | ||||
NM_001282899.1 | Intron | NP_001269828.1 | ||||
NM_001282900.1 | Intron | NP_001269829.1 | ||||
NM_001282901.1 | Intron | NP_001269830.1 | ||||
NM_007023.3 | Intron | NP_008954.2 | ||||
XM_005246246.2 | Intron | XP_005246303.1 | ||||
XM_006712205.3 | Intron | XP_006712268.1 | ||||
XM_011510505.2 | Intron | XP_011508807.1 | ||||
XM_017003196.1 | Intron | XP_016858685.1 | ||||
XM_017003197.1 | Intron | XP_016858686.1 |
RAPGEF4-AS1 - RAPGEF4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |