Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTGGGCCGGCCCCGGGAGGCT[C/T]TGAGTACCAATGAGTGCAAAGCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603051 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGPS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGPS - alkylglycerone phosphate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003659.3 | 204 | Silent Mutation | CTG,TTG | L,L 53 | NP_003650.1 | |
XM_011512041.2 | 204 | Intron | XP_011510343.1 | |||
XM_011512042.1 | 204 | Intron | XP_011510344.1 | |||
XM_011512043.2 | 204 | Intron | XP_011510345.1 |
LOC100130691 - uncharacterized LOC100130691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |