Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGTCCAAGAATCCTTTCCATCA[G/T]TGCTAAATGCTCCTTACTATCGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601951 | ||||||||||||||||||||
Literature Links: |
CLK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLK1 - CDC like kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162407.1 | 1349 | Missense Mutation | ATG,CTG | M,L 432 | NP_001155879.1 | |
NM_004071.3 | 1349 | Missense Mutation | ATG,CTG | M,L 390 | NP_004062.2 |