Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTCTTTCATAAAGAACTTCTC[A/G]TATACTTCAATATGTTCATTATTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607333 MIM: 607334 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STRADB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STRADB - STE20-related kinase adaptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206864.1 | Intron | NP_001193793.1 | ||||
NM_018571.5 | Intron | NP_061041.2 | ||||
XM_005246669.1 | Intron | XP_005246726.1 | ||||
XM_017004438.1 | Intron | XP_016859927.1 |
TRAK2 - trafficking kinesin protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |