Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGTCAGTGAGTAATGAGAAACC[A/C]TGATGCCCTACCAATTGGACTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALS2CR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALS2CR11 - amyotrophic lateral sclerosis 2 chromosome region candidate 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM237 - transmembrane protein 237 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001044385.2 | 4001 | UTR 3 | NP_001037850.1 | |||
NM_152388.3 | 4001 | UTR 3 | NP_689601.2 |