Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCATCTTTGGTCTTTGACCTTT[C/G]ACAGTTTATAATGTGCCTCAAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDBF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZDBF2 - zinc finger DBF-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285549.1 | Intron | NP_001272478.1 | ||||
NM_020923.2 | Intron | NP_065974.1 | ||||
XM_005246711.3 | Intron | XP_005246768.1 | ||||
XM_005246712.4 | Intron | XP_005246769.1 | ||||
XM_005246713.3 | Intron | XP_005246770.1 | ||||
XM_006712656.3 | Intron | XP_006712719.1 | ||||
XM_011511532.2 | Intron | XP_011509834.1 | ||||
XM_011511533.2 | Intron | XP_011509835.1 | ||||
XM_011511534.2 | Intron | XP_011509836.1 | ||||
XM_011511535.2 | Intron | XP_011509837.1 | ||||
XM_017004573.1 | Intron | XP_016860062.1 | ||||
XM_017004574.1 | Intron | XP_016860063.1 | ||||
XM_017004575.1 | Intron | XP_016860064.1 | ||||
XM_017004576.1 | Intron | XP_016860065.1 |